Learn More
Electron Microscopy Sciences Gilder Single Slot Oval Grid G153, Nickel, 1500 x 300 µm Slot
SDP
Catalog No. 501901270
Encompass_Preferred
Single Slot TEM Grids with oval holes have a thickness of are 50 microns. Each grid is individually inspected. Aperture slot is 1500x300 um. Nickel.
Material: Nickel
Slot: 1500 x 300 µm
Catalog No. 50-190-1270 Supplier Electron Microscopy Sciences Supplier No. G153NI
Single Slot TEM Grids with oval holes have a thickness of are 50 microns. Each grid is individually inspected. Aperture slot is 1500x300 um. Nickel.
Material: Nickel
Slot: 1500 x 300 µm