Learn More
Electron Microscopy Sciences Gilder Single Slot Oval Grid G215, Copper, 2000 x 150 µm Slot
SDP
Catalog No. 501901275
Encompass_Preferred
Single Slot TEM Grids with oval holes have a thickness of are 50 microns. Each grid is individually inspected.
Material: Copper
Slot: 2000 x 150 µm
Catalog No. 50-190-1275 Supplier Electron Microscopy Sciences Supplier No. G215CU
Single Slot TEM Grids with oval holes have a thickness of are 50 microns. Each grid is individually inspected.
Material: Copper
Slot: 2000 x 150 µm