Learn More
Electron Microscopy Sciences Gilder Single Slot Oval Grid GS2x1, Gold, 2000 x 1000 µm Slot
SDP
Catalog No. 501901265
Encompass_Preferred
Single Slot TEM Grids with oval holes have a thickness of are 50 microns. Each grid is individually inspected.
Material: Gold
Slot: 2000 x 1000 µm
Catalog No. 50-190-1265 Supplier Electron Microscopy Sciences Supplier No. G2010AU
Single Slot TEM Grids with oval holes have a thickness of are 50 microns. Each grid is individually inspected.
Material: Gold
Slot: 2000 x 1000 µm