Learn More
Electron Microscopy Sciences Veco Slotted Pattern Grid 50/75, Copper
SDP
Catalog No. 50997439
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter .
Material: Copper
Pitch: 500/333 µm
Hole: 450/283 µm
Bar: 50 µm
Catalog No. 50-997-439 Supplier Electron Microscopy Sciences Supplier No. 575CU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter .
Material: Copper
Pitch: 500/333 µm
Hole: 450/283 µm
Bar: 50 µm