Learn More
Electron Microscopy Sciences Veco Slotted Pattern Grid 50/75, Nickel
SDP
Catalog No. 50997440
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter .
Material: Nickel
Pitch: 500/333 µm
Hole: 450/283 µm
Bar: 50 µm
Catalog No. 50-997-440 Supplier Electron Microscopy Sciences Supplier No. 575NI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter .
Material: Nickel
Pitch: 500/333 µm
Hole: 450/283 µm
Bar: 50 µm