Learn More
Electron Microscopy Sciences Veco Slotted Pattern Grid 75/300, Nickel
SDP
Catalog No. 5028724
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter .
Material: Nickel
Pitch: 300/83 µm
Hole: 293/43 µm
Bar: 40 µm
Catalog No. 50-287-24 Supplier Electron Microscopy Sciences Supplier No. 753NI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter .
Material: Nickel
Pitch: 300/83 µm
Hole: 293/43 µm
Bar: 40 µm