Learn More
Electron Microscopy Sciences Veco Slotted Pattern Grid 100/400, Nickel
SDP
Catalog No. 50980176
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter .
Material: Nickel
Pitch: 250/63 µm
Hole: 212/25 µm
Bar: 38 µm
Catalog No. 50-980-176 Supplier Electron Microscopy Sciences Supplier No. 1040NI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter .
Material: Nickel
Pitch: 250/63 µm
Hole: 212/25 µm
Bar: 38 µm