Learn More
Electron Microscopy Sciences Easy-Load Slotted Tissue Cassettes, Green
SDP
Catalog No. 5030295
Encompass_Preferred
These cassettes made from acetal copolymer allow you to load the Micro-Writer and Shur/Mark Cassette Labeling Instruments in one simple operation. Each convenient stack contains 50 unique cassettes. No more manual insertion one-by-one.
The anterior printing area is a 45 degree angle which suits most cassette labeling instruments. One-piece disposable plastic lids are available separately. The lids can be opened and closed as often as necessary and it always relock securely without danger of specimen loss.
Cassettes are packaged in stacks of 50. Each case contains 2 boxes of 20 stacks.
Color: Green
Catalog No. 50-302-95 Supplier Electron Microscopy Sciences Supplier No. 70060G
These cassettes made from acetal copolymer allow you to load the Micro-Writer and Shur/Mark Cassette Labeling Instruments in one simple operation. Each convenient stack contains 50 unique cassettes. No more manual insertion one-by-one.
The anterior printing area is a 45 degree angle which suits most cassette labeling instruments. One-piece disposable plastic lids are available separately. The lids can be opened and closed as often as necessary and it always relock securely without danger of specimen loss.
Cassettes are packaged in stacks of 50. Each case contains 2 boxes of 20 stacks.
Color: Green