Learn More
Bellco Glass, Inc. Slow Speed Kit, .085-1.71 RPM
SDP
Catalog No. 501945906
Encompass_Preferred
The slow speed conversion kits adapt any modular or fixed frame cell production roller apparatus to operate between either the speeds of 0.10 and 2.0 rpm (7630-60666) or the speeds of .085-1.7 rpm (7630-70777). MADE IN THE USA
Catalog No. 50-194-5906 Supplier Bellco Glass, Inc. Supplier No. 763070777
The slow speed conversion kits adapt any modular or fixed frame cell production roller apparatus to operate between either the speeds of 0.10 and 2.0 rpm (7630-60666) or the speeds of .085-1.7 rpm (7630-70777). MADE IN THE USA