Learn More
Bellco Glass, Inc. Slow Speed Kit, .10-2.0 RPm
SDP
Catalog No. 501945905
Encompass_Preferred
The slow speed conversion kits adapt any modular or fixed frame cell production roller apparatus to operate between either the speeds of 0.10 and 2.0 rpm (7630-60666) or the speeds of .10-2.0 rpm (7630-70777). MADE IN THE USA
Catalog No. 50-194-5905 Supplier Bellco Glass, Inc. Supplier No. 763060666
The slow speed conversion kits adapt any modular or fixed frame cell production roller apparatus to operate between either the speeds of 0.10 and 2.0 rpm (7630-60666) or the speeds of .10-2.0 rpm (7630-70777). MADE IN THE USA