Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SlowFade™ Glass Soft-Set Antifade Mountant
GREENER_CHOICE

Catalog No. S36917
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
2 mL
5 x 2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S36917 2 mL
S369175X2ML 5 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S36917 Supplier Invitrogen™ Supplier No. S36917
  • $94.75
    $77.70 / Each of 1

    Save $17.05

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

SlowFade Glass Antifade Mountant is a glycerol-based, ready-to-use, non-curing mountant applied directly to fluorescently labeled cell or tissue samples on microscope slides.

  • SlowFade Glass Antifade Mountant is a glycerol-based, ready-to-use, non-curing mountant applied directly to fluorescently labeled cell or tissue samples on microscope slides. It is formulated to have a refractive index of 1.52 for sharper images and antifade components to resist photobleaching.
  • The result is sharp, bright images with almost any fluorophore, with wide-field, confocal, or super-resolution microscopes, at focal depths of 0–500 μm.
  • Since SlowFade Glass Antifade Mountant is non-curing, cells can be imaged immediately after mounting, without any of the adverse effects on cell morphology that a curing mountant might cause. It comes ready to use—just apply a drop to the sample, add a coverslip, and image. It is available with or without DAPI nuclear stain.
  • If a curing, hard-set mountant is needed, also with a refractive index of 1.52, for years of slide storage, then Prolong Glass Antifade Mountant is recommended.
  • Ready-to-use, glycerol-based, non-curing antifade mountant
  • Refractive index of 1.52, with minimized spherical aberrations
  • Up to three times better axial resolution compared to regular mountant
  • Visible tissue optical clearing, even in sections up to 1-mm thick
  • Sharp, bright images at any focal depth from 0 to 500 μm
  • SlowFade Glass Antifade Mountant features a refractive index of 1.52 (similar to glass coverslips), with superior photobleach protection. It is compatible with immersion oil and oil-immersion microscope optics, enabling a refraction-free light path and minimized spherical aberration, which improves axial resolution by three times (at 150-μm focal depth) as measured by point spread function.
  • When 1-mm thick tissue sections are mounted, there is visible optical tissue clearing, enabling deep 3D imaging. For 3D imaging of whole organs (up to 10 mm), please see Clearing Reagents for Imaging 3D Cell Culture and Tissue.
  • No matter which fluorescent dyes or fluorescent proteins you are using, Slowfade Glass Antifade Mountant helps provide superior protection against photobleaching. It has been shown to have best-in-class performance with a wide array of fluorescent dyes, including Alexa Fluor, FITC, TRTC, Texas Red, Cy3, and Cy5 dyes, and fluorescent proteins, including GFP, RFP, and mCherry.
  • SlowFade Glass mountant does not quench fluorophores, and its ability to provide brighter signal against no background helps ensure that you get the best-quality high-resolution images for publishing.
  • Most tissue clearing techniques either expand or shrink the specimens, causing artifacts in specimen morphology. SlowFade Glass Antifade Mountant is non-curing and does not cause expansion or contraction of specimens, which we have verified using Hela spheroids and rat brain sections. This characteristic allows 3D imaging of truly native specimen morphology. The specimens are available for imaging immediately after mounting, and properly sealed slides can be stored for weeks or months for excellent
TRUSTED_SUSTAINABILITY

Specifications

Product Type Antifade Mountant
Content And Storage Contains 1 dropper bottle containing 2 mL reagent
Shipping Condition Approved for shipment on Wet or Dry Ice
Product Line SlowFade
Reagent Type Antifade Solution
Volume (Metric) 2 mL
May I use nucleic acid stains for samples that will be mounted with SlowFade Glass Soft-set Antifade Mountant (Cat. No. S36917)?

Yes. SlowFade Glass Soft-set Antifade Mountant (Cat. No. S36917) is compatible with many nucleic acid stains. Apply the stain at the last step prior to mounting.

Will SlowFade Glass Soft-set Antifade Mountant (Cat. No. S36917) preserve the structure of fluorescent proteins?

No. SlowFade Glass Soft-set Antifade Mountant (Cat. No. S36917) does not maintain the structures of fluorescent proteins over time. The proteins will naturally unfold, and this unfolding would result in the loss of fluorescence.

What can be done to remove any tiny air bubbles in SlowFade Glass Soft-set Antifade Mountant (Cat. No. S36917)?

To remove tiny air bubbles, place the vial of SlowFade Glass Soft-set Antifade Mountant (Cat. No. S36917) within a vacuum chamber with the vial cap loosened. Degas the solution by applying vacuum through either a T-connector on the faucet or a very brief application of vacuum from a vacuum pump.

Does SlowFade Glass Soft-set Antifade Mountant (Cat. No. S36917) contain any volatile organic solvents?

No. SlowFade Glass Soft-set Antifade Mountant (Cat. No. S36917) is an aqueous-based mountant.

Does Slowfade Antifade Mountant require using a nail polish for sealing?

No. Samples mounted with SlowFade mountants need not be sealed; they are intended for immediate viewing. The coverslip may be anchored (to prevent movement while viewing) by applying molten paraffin to three or four spots around the edge of the coverslip.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.