Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Slp7111228 hydrochloride | 1449768-48-2 | 98.0% | 5 MG
SDP

Catalog No. 5000233414
Encompass_Preferred

SLP7111228 hydrochloride is the hydrochloride salt of SLP7111228 supplied as a solid research reagent for biochemical assay use. It has formula C22H34ClN5O, molecular weight 419.99, and CAS 1449768-48-2. Store sealed at -20°C; in solution store at -80°C for long-term stability.

  • Hydrochloride salt form.
  • Solid, white/off-white appearance.
  • Molecular formula C22H34ClN5O.
  • Molecular weight 419.99.
  • CAS number 1449768-48-2.
  • Storage: -20°C; in solvent -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-33414 Supplier Medchemexpress LLC Supplier No. HY120152A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SLP7111228 hydrochloride is the hydrochloride salt of SLP7111228 supplied as a solid research reagent for biochemical assay use. It has formula C22H34ClN5O, molecular weight 419.99, and CAS 1449768-48-2. Store sealed at -20°C; in solution store at -80°C for long-term stability.

  • Hydrochloride salt form.
  • Solid, white/off-white appearance.
  • Molecular formula C22H34ClN5O.
  • Molecular weight 419.99.
  • CAS number 1449768-48-2.
  • Storage: -20°C; in solvent -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.