Learn More
Cayman Chemical SLP7111228hydrochlorIde 10mg
SDP
Catalog No. 502993774
Encompass_Preferred
A potent and selective SPHK1 inhibitor (Kis = 0.048 and >10 μM for SPHK1 and 2, respectively); induces a concentration-dependent decrease in U937 cells; decreases blood levels of S1P in a dose-dependent manner in rats
Catalog No. 50-299-3774 Supplier Cayman Chemical Supplier No. 2329010 MG
A potent and selective SPHK1 inhibitor (Kis = 0.048 and >10 μM for SPHK1 and 2, respectively); induces a concentration-dependent decrease in U937 cells; decreases blood levels of S1P in a dose-dependent manner in rats