Learn More
Enzo Life Sciences SLPI (human), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI9002220001
Encompass_Preferred
The SLPI ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of SLPI in serum with results in just 2.5 hours. Alternative name: Secretory leukocyte proteinase inhibitor, ALP, Antileukoproteinase. Assay Time: 2.5 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human .
Catalog No. 50-200-6874