Learn More
SPEX CERTIPREP LLC 3P 48 Component ICP-MS Standard at 10 ug/mL Solution A, 100 mL
Catalog No. NC0889966
Encompass_Preferred
(HPS by Zeptometrix) ICP-MS Multi Element Solution Standard with 48 Components at 10 ug/mL. This is Solution A of the 3 Solution Set ICP-MS-68A.
Catalog No. NC0889966 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICPMS68AA100
(HPS by Zeptometrix) ICP-MS Multi Element Solution Standard with 48 Components at 10 ug/mL. This is Solution A of the 3 Solution Set ICP-MS-68A.