Learn More
Medchemexpress LLC SLU-PP-1072 | 2285432-57-5 | C18H12N2O3S | 25 MG
SDP
Catalog No. 5000375564
Encompass_Preferred
SLU-PP-1072 is a dual ERRα/γ inverse agonist identified for its role in prostate cancer (PCa) research. This compound functions by disrupting PCa cell metabolism and inducing apoptosis through the dysregulation of the cell cycle. It acts as a selective inverse agonist for ERRα and ERRγ, making it a preclinical tool for targeting ERRα/γ-driven cancer metabolism.
  • Acts as a dual ERRα/γ inverse agonist.
  • Used in prostate cancer (PCa) research.
  • Disrupts PCa cell metabolism.
  • Induces apoptosis by dysregulating the cell cycle.
  • Selective for ERRα/γ.
  • Inhibits the Warburg effect.
Catalog No. 50-003-75564 Supplier Medchemexpress LLC Supplier No. 25MGHY153895
SLU-PP-1072 is a dual ERRα/γ inverse agonist identified for its role in prostate cancer (PCa) research. This compound functions by disrupting PCa cell metabolism and inducing apoptosis through the dysregulation of the cell cycle. It acts as a selective inverse agonist for ERRα and ERRγ, making it a preclinical tool for targeting ERRα/γ-driven cancer metabolism.
  • Acts as a dual ERRα/γ inverse agonist.
  • Used in prostate cancer (PCa) research.
  • Disrupts PCa cell metabolism.
  • Induces apoptosis by dysregulating the cell cycle.
  • Selective for ERRα/γ.
  • Inhibits the Warburg effect.