Learn More
Medchemexpress LLC SLU-PP-332 /25MG
SDP
Catalog No. NC2739531
Encompass_Preferred
SLU-PP-332 is a pan-Estrogen Receptor/ERR agonist with EC50 values of 98 / 230 / and 430 nM. Respectively SLUPP-332 enhances mitochondrial function and cellular respiration in skeletal muscle cell lines. For research use only.
Catalog No. NC2739531 Supplier Medchemexpress LLC Supplier No. HY15567325MG
SLU-PP-332 is a pan-Estrogen Receptor/ERR agonist with EC50 values of 98 / 230 / and 430 nM. Respectively SLUPP-332 enhances mitochondrial function and cellular respiration in skeletal muscle cell lines. For research use only.