Learn More
Aviva Systems Biology Corp Slu7 Antibody - middle region (ARP52180_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52180_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Slu7. It was validated on Western Blot. . Alternate names: AU018913, D11Ertd730e, D3Bwg0878e, MGC31026, RP23-26M1.8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-656