Learn More
Cole-Parmer Raven Environmental SID-10200-30 Environmental Products Sludge Interface Detector with 30 Foot Cable
Supplier:  Cole-Parmer 0595620
Encompass_Preferred
The SID-10200 is a portable handheld instrument that accurately determines the sludge blanket solid/liquid interface depths in primary tanks, clarifiers, sedimentation basins and a wide variety of sludge settling tanks. The solid/liquid interface is detected when the probe is lowered into the tank. As solids density increses the meter's LED lightbars begin to illuminate and the alarm horn frequency increases. The meter has a sensor with a 30-ft cable that has 1-ft incremented depth marks. The unit is powered by six AA batteries (included) and measures 8"L x 4.5"W x 3.5"D
Catalog No. 50-632-419