Learn More
Glass Expansion Slurry DC Nebulizer 4mL/min with 1,500mm tubing, Includes NFTS-075 UniFit Connector with 0.75mm ID x 1.3mm OD x 1500mm long tubing (1 piece), and 70-803-1311 Gas Fitting with Ratchet Nebulizer connector
SDP
Catalog No. 501907487
Encompass_Preferred
Slurry DC Nebulizer 4.0mL/min, includes UniFit Connector with 0.75mm ID x 1.3mm OD x 700mm long tubing (1 pc) and DC Gas Fitting Type 41 with ratchet connector (1 pc). Compatible with Shimadzu 9000/9800 ICP-OES.Optimum nebulizer gas flow rate is 1.0L/min and sample flow rate is between 1.5 and 2.5mL/min. Recommended not to exceed 4.0mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read approximately 40 psi at 1.0L/min gas flow rate. The Slurry nebulizer is ideal for aspirating slurries or suspensions as it has a high tolerance for particulates, up to 150um. TDS tolerance up to 1%.Replacement parts include P/Ns NC0277960 and 501907301.Recommend nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7487 Supplier Glass Expansion Supplier No. A411US6S
Slurry DC Nebulizer 4.0mL/min, includes UniFit Connector with 0.75mm ID x 1.3mm OD x 700mm long tubing (1 pc) and DC Gas Fitting Type 41 with ratchet connector (1 pc). Compatible with Shimadzu 9000/9800 ICP-OES.Optimum nebulizer gas flow rate is 1.0L/min and sample flow rate is between 1.5 and 2.5mL/min. Recommended not to exceed 4.0mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read approximately 40 psi at 1.0L/min gas flow rate. The Slurry nebulizer is ideal for aspirating slurries or suspensions as it has a high tolerance for particulates, up to 150um. TDS tolerance up to 1%.Replacement parts include P/Ns NC0277960 and 501907301.Recommend nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001