Learn More
Perkin Elmer US LLC Slurry Glass Nebulizer, 4.0 mL/min, for Optima 3x00 RL
Catalog No. 502104656
Encompass_Preferred
A sample which consists of small particulates in liquid matrix, known as a Slurry, is suited for this nebulizer. Slurry nebulizers have a natural liquid uptake of 4 mL/min but operate best between 1.5 - 2.5 mL/min
Catalog No. 50-210-4656 Supplier Perkin Elmer US LLC Supplier No. N8121059
A sample which consists of small particulates in liquid matrix, known as a Slurry, is suited for this nebulizer. Slurry nebulizers have a natural liquid uptake of 4 mL/min but operate best between 1.5 - 2.5 mL/min