Learn More
Cayman Chemical (±)-SLV 319, Purity: ≥98%, CAS: 362519-49-1, 1 mg
SDP
Catalog No. 502888857
Encompass_Preferred
A mixture of the potent and selective CB1 receptor antagonist SLV 319 (Ki = 7.8 nM) and its distomer, SLV 319 (+)-enantiomer
Catalog No. 50-288-8857 Supplier Cayman Chemical Supplier No. 100092261 MG
A mixture of the potent and selective CB1 receptor antagonist SLV 319 (Ki = 7.8 nM) and its distomer, SLV 319 (+)-enantiomer