Learn More
MSC Daubert Cromwell Silver Saver Bag, 6 x 8 in, 250/Pk
Catalog No. 501917338
Encompass
The Silver Saver bag is designed to protect silver, gold, and other noble metals from tarnish and corrosion. It is constructed from bleached kraft paper coated with a special chemical formulation that absorbs hydrogen sulfide and other environmental elements, providing effective protection for up to two years.
  • Protects silver, gold, and noble metals from tarnish
  • Absorbs hydrogen sulfide and other corrosive elements
  • Provides up to two years of protection
  • Constructed from bleached kraft with a chemical coating
  • Ideal for shipping and packaging applications
  • White color
  • Dimensions: 6 in width, 8 in height
  • Packaged as 250 bags per case
Catalog No. 50-191-7338 Supplier MSC Supplier No. 39569009
The Silver Saver bag is designed to protect silver, gold, and other noble metals from tarnish and corrosion. It is constructed from bleached kraft paper coated with a special chemical formulation that absorbs hydrogen sulfide and other environmental elements, providing effective protection for up to two years.
  • Protects silver, gold, and noble metals from tarnish
  • Absorbs hydrogen sulfide and other corrosive elements
  • Provides up to two years of protection
  • Constructed from bleached kraft with a chemical coating
  • Ideal for shipping and packaging applications
  • White color
  • Dimensions: 6 in width, 8 in height
  • Packaged as 250 bags per case