Learn More
Cayman Chemical Slydnn 10mg
SDP
Catalog No. 502918514
Encompass_Preferred
A flavonolignan with diverse biological activities; inhibits tyrosinase (IC50 = 2.6 µM for the mushroom enzyme); scavenges DPPH radicals in a cell-free assay (IC50 = 66.8 µM); protects against allyl alcohol-, carbon tetrachloride-, paracetamol-, or D-galactosamine-induced cytotoxicity in primary human hepatocytes at 100 µM; inhibits the proliferation of DU145 and PC3 cells at 90 µM; prevents methacholine-induced AHR and reduces blood and BALF levels of eosinophils in an ovalbumin-sensitized mouse model of allergic asthma when administered via inhalation at 1%
Catalog No. 50-291-8514 Supplier Cayman Chemical Supplier No. 3924810 MG
A flavonolignan with diverse biological activities; inhibits tyrosinase (IC50 = 2.6 µM for the mushroom enzyme); scavenges DPPH radicals in a cell-free assay (IC50 = 66.8 µM); protects against allyl alcohol-, carbon tetrachloride-, paracetamol-, or D-galactosamine-induced cytotoxicity in primary human hepatocytes at 100 µM; inhibits the proliferation of DU145 and PC3 cells at 90 µM; prevents methacholine-induced AHR and reduces blood and BALF levels of eosinophils in an ovalbumin-sensitized mouse model of allergic asthma when administered via inhalation at 1%