Learn More
Apexbio Technology LLC SM-102 100mg
SDP
Catalog No. 503107262
Encompass_Preferred
SM-102 is an amino cationic lipid specifically used to form lipid nanoparticles (LNPs) to enhance the efficiency of mRNA delivery into cells Studies have shown that SM-102 effectively modulates erg-mediated potassium currents (i erg) in GH cells at concentrations ranging from 100 to 300 M thereby regulating specific signaling pathways Its applications include research on drug delivery technologies for mRNA therapies and vaccine development
Catalog No. 50-310-7262 Supplier Apexbio Technology LLC Supplier No. C1042250
SM-102 is an amino cationic lipid specifically used to form lipid nanoparticles (LNPs) to enhance the efficiency of mRNA delivery into cells Studies have shown that SM-102 effectively modulates erg-mediated potassium currents (i erg) in GH cells at concentrations ranging from 100 to 300 M thereby regulating specific signaling pathways Its applications include research on drug delivery technologies for mRNA therapies and vaccine development