Learn More
Medchemexpress LLC HY-111482 10mg Medchemexpress, SM 16 CAS:614749-78-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11148210MG
Encompass_Preferred
Medchemexpress, HY-111482 10mg SM 16 CAS:614749-78-9 SM 16 is a ALK5/ALK4 kinase inhibitor with Kis of 10 and 1.5 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8879