Learn More
Apexbio Technology LLC SM-164, 10mg. Cas: 957135-43-2 MFCD: MFCD28168019. Anticancer agent
SDP
Supplier:  Apexbio Technology LLC A881510
Encompass_Preferred
SM-164 is a bivalent mimetic of Smac with Ki values of 0.31 nM, 1.1 nM and 0.56 nM for cIAP-1, cIAP-2 and XIAP, respectively 1.SM-164 is developed as an anticancer agent. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5194