Learn More
Medchemexpress LLC HY-15989 10mg , SM-164 CAS:957135-43-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1598910MG
Encompass_Preferred
HY-15989 10mg SM-164 CAS: 957135-43-2 SM-164 is a cell-permeable Smac mimetic compound. SM-164 binds to XIAP protein containing both the BIR2 and BIR3 domains with an IC50 value of 1.39 nM and functions as an extremely potent antagonist of XIAP. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9543