Learn More
Selleck Chemical LLC SM-164 5mg
SDP
Supplier:  Selleck Chemical LLC S70895MG
Encompass_Preferred
SM-164 is a potent, non-peptide, cell-permeable antagonist of XIAP that targets both the BIR2 and BIR3 domains with IC50 of 1.39 nM. SM-164 induces apoptosis and tumor regression. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3253