Learn More
TARGETMOL CHEMICALS INC SM-164 5MG
SDP
Catalog No. 502581394
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. SM-164 binds to XIAP protein containing both the BIR2 and BIR3 domains (IC50 1.39 nM). It acts as an extremely potent antagonist of XIAP. purity: 99%
Catalog No. 50-258-1394 Supplier TARGETMOL CHEMICALS INC Supplier No. T12932L5MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. SM-164 binds to XIAP protein containing both the BIR2 and BIR3 domains (IC50 1.39 nM). It acts as an extremely potent antagonist of XIAP. purity: 99%