Learn More
Apexbio Technology LLC SM-164, 957135-43-2, MFCD28167763, 5 mg
SDP
Catalog No. 501015392
Encompass_Preferred
MF: C62H84N14O6, MW: 1121.42, Solubility: 56.071mg/mL in DMSO. SM-164 is a bivalent mimetic of Smac with Ki values of 0.31 nM, 1.1 nM and 0.56 nM for cIAP-1, cIAP-2 and XIAP, respectively.
Catalog No. 50-101-5392 Supplier Apexbio Technology LLC Supplier No. A88155
MF: C62H84N14O6, MW: 1121.42, Solubility: 56.071mg/mL in DMSO. SM-164 is a bivalent mimetic of Smac with Ki values of 0.31 nM, 1.1 nM and 0.56 nM for cIAP-1, cIAP-2 and XIAP, respectively.