Learn More
Medchemexpress LLC HY-15989A 5mg Medchemexpress, SM-164 Hydrochloride CAS: Purity:>98%
SDP
Catalog No. 501969965
Encompass_Preferred
Medchemexpress, HY-15989A 5mg SM-164 HCl CAS: SM-164 Hydrochloride is a cell-permeable Smac mimetic compound. SM-164 binds to XIAP protein containing both the BIR2 and BIR3 domains with an IC50 value of 1.39 nM and functions as an extremely potent antagonist of XIAP. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9965 Supplier Medchemexpress LLC Supplier No. HY15989A5MG
Medchemexpress, HY-15989A 5mg SM-164 HCl CAS: SM-164 Hydrochloride is a cell-permeable Smac mimetic compound. SM-164 binds to XIAP protein containing both the BIR2 and BIR3 domains with an IC50 value of 1.39 nM and functions as an extremely potent antagonist of XIAP. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.