Learn More
Medchemexpress LLC Sm-360320 | 226907-52-4 | 99.4% | 315.33 g/mol | C15H17N5O3 | 100MG
SDP
Catalog No. 5000235421
Encompass_Preferred
SM-360320 (CL-087) is a small-molecule Toll-like receptor 7 (TLR7) agonist used in preclinical research as an immunomodulator with reported antitumor and antiviral activity. It is supplied as a solid with high purity and characterized potency for cell-based and immunological assays.
  • Potent TLR7 agonist with documented EC50 values in HEK293 assays (1.17 μM and 399.4 nM).
  • Shows antiviral activity against HCV in Huh7 cells (EC50 30 nM).
  • Induces IFNα production in human PBMCs (EC50 0.14 μM).
  • High purity solid (99.41%) suitable for preclinical studies.
  • Storage recommendations: powder at -20°C for long-term storage; in solvent at -80°C for up to 6 months.
Catalog No. 50-002-35421 Supplier Medchemexpress LLC Supplier No. HY125390100MG
SM-360320 (CL-087) is a small-molecule Toll-like receptor 7 (TLR7) agonist used in preclinical research as an immunomodulator with reported antitumor and antiviral activity. It is supplied as a solid with high purity and characterized potency for cell-based and immunological assays.
  • Potent TLR7 agonist with documented EC50 values in HEK293 assays (1.17 μM and 399.4 nM).
  • Shows antiviral activity against HCV in Huh7 cells (EC50 30 nM).
  • Induces IFNα production in human PBMCs (EC50 0.14 μM).
  • High purity solid (99.41%) suitable for preclinical studies.
  • Storage recommendations: powder at -20°C for long-term storage; in solvent at -80°C for up to 6 months.