Learn More
Medchemexpress LLC Sm-360320 (CL-087) | 226907-52-4 | 99.4% | 315.33 | C15H17N5O3 | 1MG
SDP
Catalog No. 5000239317
Encompass_Preferred
SM-360320 (CL-087) is a small-molecule Toll-like receptor 7 (TLR7) agonist used for research. It functions as an immunomodulator with reported antitumor activity, can enhance Th1 antibody responses when combined with DNA vaccines, and has been reported to inhibit hepatitis C virus replication via both interferon-mediated and interferon-independent mechanisms.
  • Potent TLR7 agonist for immune modulation.
  • Reported antitumor activity in preclinical studies.
  • Enhances Th1 antibody responses with DNA vaccines.
  • Inhibits HCV replication through IFN-dependent and IFN-independent mechanisms.
  • Available in solid and solution formats with multiple pack sizes.
  • High reported purity suitable for research applications.
Catalog No. 50-002-39317 Supplier Medchemexpress LLC Supplier No. HY1253901MG
SM-360320 (CL-087) is a small-molecule Toll-like receptor 7 (TLR7) agonist used for research. It functions as an immunomodulator with reported antitumor activity, can enhance Th1 antibody responses when combined with DNA vaccines, and has been reported to inhibit hepatitis C virus replication via both interferon-mediated and interferon-independent mechanisms.
  • Potent TLR7 agonist for immune modulation.
  • Reported antitumor activity in preclinical studies.
  • Enhances Th1 antibody responses with DNA vaccines.
  • Inhibits HCV replication through IFN-dependent and IFN-independent mechanisms.
  • Available in solid and solution formats with multiple pack sizes.
  • High reported purity suitable for research applications.