Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sm-360320 (synonym CL-087) | 226907-52-4 | 99.4% | 315.33 g/mol | C15H17N5O3 | 10MG
SDP

Catalog No. 5000238966
Encompass_Preferred

SM-360320 is a high-purity small-molecule Toll-like receptor 7 (TLR7) agonist provided for research applications in immunology and cancer-immunotherapy studies. It is supplied in powder and solution formats, with supporting documentation for experimental use. The compound is intended for laboratory research only and is not for clinical use.

  • High purity: 99.4% (manufacturer-reported)
  • Chemical formula: C15H17N5O3; molecular weight: 315.33 g/mol
  • CAS number: 226907-52-4 for unambiguous identification
  • Available in multiple pack sizes and as a 10 mM solution in DMSO
  • Storage: powder at -20°C; in solvent at -80°C (long-term) or -20°C (short-term)
  • Suitable for in vitro and in vivo immunomodulation and screening assays
  • Documentation available: datasheet, certificate of analysis, and safety data sheet

Catalog No. 50-002-38966 Supplier Medchemexpress LLC Supplier No. HY12539010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SM-360320 is a high-purity small-molecule Toll-like receptor 7 (TLR7) agonist provided for research applications in immunology and cancer-immunotherapy studies. It is supplied in powder and solution formats, with supporting documentation for experimental use. The compound is intended for laboratory research only and is not for clinical use.

  • High purity: 99.4% (manufacturer-reported)
  • Chemical formula: C15H17N5O3; molecular weight: 315.33 g/mol
  • CAS number: 226907-52-4 for unambiguous identification
  • Available in multiple pack sizes and as a 10 mM solution in DMSO
  • Storage: powder at -20°C; in solvent at -80°C (long-term) or -20°C (short-term)
  • Suitable for in vitro and in vivo immunomodulation and screening assays
  • Documentation available: datasheet, certificate of analysis, and safety data sheet

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.