Learn More
Medchemexpress LLC Sm-433 | 1071992-81-8 | 98.8% | C32H43N5O4 | 10MG
SDP
Catalog No. 5000260448
Encompass_Preferred
SM-433 is a Smac mimetic small-molecule research reagent that targets inhibitor of apoptosis proteins (IAPs) by binding the XIAP BIR3 domain (reported IC50 <1 μM). It is provided as a high-purity compound for biochemical and cellular studies of apoptosis signaling and IAP-targeted mechanisms.
  • Binds XIAP BIR3 with reported IC50 <1 μM.
  • Suitable for biochemical and cellular assays investigating apoptosis.
  • High reported purity (~98.8%).
  • Soluble in DMSO for assay preparation.
  • Provided in small milligram quantities for research use.
Catalog No. 50-002-60448 Supplier Medchemexpress LLC Supplier No. HY13805910MG
SM-433 is a Smac mimetic small-molecule research reagent that targets inhibitor of apoptosis proteins (IAPs) by binding the XIAP BIR3 domain (reported IC50 <1 μM). It is provided as a high-purity compound for biochemical and cellular studies of apoptosis signaling and IAP-targeted mechanisms.
  • Binds XIAP BIR3 with reported IC50 <1 μM.
  • Suitable for biochemical and cellular assays investigating apoptosis.
  • High reported purity (~98.8%).
  • Soluble in DMSO for assay preparation.
  • Provided in small milligram quantities for research use.