Learn More
Medchemexpress LLC SM-433 hydrochloride | 98.6% | 598.18 | 25 MG
SDP
Catalog No. 5000351038
Encompass_Preferred
SM-433 hydrochloride is a Smac mimetic that functions as an inhibitor of inhibitor of apoptosis proteins (IAPs). It demonstrates strong binding affinity to the XIAP BIR3 protein with an IC50 of less than 1 μM. This product is for research use only and not for sale to patients.
  • Smac mimetic
  • Inhibitor of inhibitor of apoptosis proteins (IAPs)
  • Strong binding affinity to XIAP BIR3 protein (IC50 < 1 μM)
  • Exhibits strong inhibitory activity against MDA-MB -2131 human breast cancer cells and SK-OV-3 ovarian cancer cells (IC50s < 10 μM) in vitro
Catalog No. 50-003-51038 Supplier Medchemexpress LLC Supplier No. 25MGHY138059A
SM-433 hydrochloride is a Smac mimetic that functions as an inhibitor of inhibitor of apoptosis proteins (IAPs). It demonstrates strong binding affinity to the XIAP BIR3 protein with an IC50 of less than 1 μM. This product is for research use only and not for sale to patients.
  • Smac mimetic
  • Inhibitor of inhibitor of apoptosis proteins (IAPs)
  • Strong binding affinity to XIAP BIR3 protein (IC50 < 1 μM)
  • Exhibits strong inhibitory activity against MDA-MB -2131 human breast cancer cells and SK-OV-3 ovarian cancer cells (IC50s < 10 μM) in vitro