Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SM-433, a Smac mimetic | 1071992-81-8 | 99.0% | 561.71 g/mol | C32H43N5O4 | 5 MG
SDP

Catalog No. 5000222711
Encompass_Preferred

SM-433 is a Smac mimetic small molecule that inhibits inhibitor of apoptosis proteins (IAPs), with reported binding to the XIAP BIR3 domain and cellular activity in cancer cell lines. Supplied as a solid for research use.

  • Functions as a Smac mimetic and IAP inhibitor.
  • Binds XIAP BIR3 with reported IC50 ~1 μM.
  • Shows cellular activity (IC50s <10 μM) in select cancer cell lines.
  • Chemical formula C32H43N5O4; molecular weight 561.71 g/mol.
  • High purity (~99%) and supplied as a white to off-white solid.
  • Soluble in DMSO (160 mg/mL); in vivo formulation protocols available.
  • Storage: keep sealed at 4°C; in solvent store at -80°C long term.

Catalog No. 50-002-22711 Supplier Medchemexpress LLC Supplier No. HY1380595MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SM-433 is a Smac mimetic small molecule that inhibits inhibitor of apoptosis proteins (IAPs), with reported binding to the XIAP BIR3 domain and cellular activity in cancer cell lines. Supplied as a solid for research use.

  • Functions as a Smac mimetic and IAP inhibitor.
  • Binds XIAP BIR3 with reported IC50 ~1 μM.
  • Shows cellular activity (IC50s <10 μM) in select cancer cell lines.
  • Chemical formula C32H43N5O4; molecular weight 561.71 g/mol.
  • High purity (~99%) and supplied as a white to off-white solid.
  • Soluble in DMSO (160 mg/mL); in vivo formulation protocols available.
  • Storage: keep sealed at 4°C; in solvent store at -80°C long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.