Learn More
Promocell GmbH PromoCell Small Airway Epithelial Cell Growth Medium KIT 500 ml, Contains: C-21270, C-39170
Catalog No. 50306181
Encompass
PromoCell Small Airway Epithelial Cell Growth Medium is a serum-free medium developed for the cultivation of epithelial cells from the distal respiratory system. The optimal adjustment of growth factors in this medium ensures excellent in vitro growth.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for porcine small airway epithelial cells.| Bovine Pituitary Extract | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Insulin (recombinant human) | 5 μg / ml || Hydrocortisone | 0.5 μg / ml || Epinephrine | 0.5 μg / ml || Triiodo-L-thyronine | 6.7 ng / ml || Transferrin (recombinant human) | 10 μg / ml || Retinoic Acid | 0.1 ng / ml || Bovine Serum Albumin-Fatty Acid Free (BSA-FAF) | 2.5 mg/ml |
Catalog No. 50-306-181 Supplier Promocell GmbH Supplier No. C21170
PromoCell Small Airway Epithelial Cell Growth Medium is a serum-free medium developed for the cultivation of epithelial cells from the distal respiratory system. The optimal adjustment of growth factors in this medium ensures excellent in vitro growth.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for porcine small airway epithelial cells.| Bovine Pituitary Extract | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Insulin (recombinant human) | 5 μg / ml || Hydrocortisone | 0.5 μg / ml || Epinephrine | 0.5 μg / ml || Triiodo-L-thyronine | 6.7 ng / ml || Transferrin (recombinant human) | 10 μg / ml || Retinoic Acid | 0.1 ng / ml || Bovine Serum Albumin-Fatty Acid Free (BSA-FAF) | 2.5 mg/ml |