Learn More
Medicus Health Splash Shield Benchtop BioHazard, Small
SDP
Catalog No. NC3371774
Encompass
Benchtop Biohazard Safety Splash Shield increases splash protection by keeping hazardous material contained while you work
∙ Durable, scratch-resistant plexiglass shield protects you from splashes, aerosol mist, and flying debris
∙ Economic alternative provides great splash protection
Helps meet compliance with OSHA bloodborne pathogen regulations (29 CFR Part 191.1030)
∙ Lab Shields are made from durable PETG plastic that's 20 times stronger than acrylic substitute shields which crack, splinter, craze and yellow easily - accept no substitutes
∙ Clear shield provides easy viewing and can be cleaned with any standard laboratory disinfectant
∙ A nice addition to a fume hood
∙ Skid resistant feet keep shield in place and make it easy to position
∙ Dimensions: 11.5"W x 7"D x 15"H
Catalog No. NC3371774 Supplier Medicus Health Supplier No. 2484M1
Benchtop Biohazard Safety Splash Shield increases splash protection by keeping hazardous material contained while you work
∙ Durable, scratch-resistant plexiglass shield protects you from splashes, aerosol mist, and flying debris
∙ Economic alternative provides great splash protection
Helps meet compliance with OSHA bloodborne pathogen regulations (29 CFR Part 191.1030)
∙ Lab Shields are made from durable PETG plastic that's 20 times stronger than acrylic substitute shields which crack, splinter, craze and yellow easily - accept no substitutes
∙ Clear shield provides easy viewing and can be cleaned with any standard laboratory disinfectant
∙ A nice addition to a fume hood
∙ Skid resistant feet keep shield in place and make it easy to position
∙ Dimensions: 11.5"W x 7"D x 15"H