Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SM Buffer

Catalog No. 113070044 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-307-0044 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-307-0044 Supplier MP Biomedicals™ Supplier No. 113070044
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SM Buffer is a sterile solution.

  • Sterile Solution
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Physical Form Solution
Quantity 500 mL
Solution Type Buffer
Sterility Sterile
Solubility Information Insoluble
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.