Learn More
Teknova SM Buffer. 1000mL, Sterile 2 pack.
SDP
Catalog No. 502387302
Encompass
100mM Sodium Chloride, 8mM Magnesium Sulfate,0.01% Gelatin, 50mM Tris-HCl
Catalog No. 50-238-7302 Supplier Teknova Supplier No. S22102PK
100mM Sodium Chloride, 8mM Magnesium Sulfate,0.01% Gelatin, 50mM Tris-HCl