Learn More
Electron Microscopy Sciences Small FIB Grid Holder, Pin, 12.7 mm
SDP
Catalog No. 501927127
Encompass_Preferred
Holds up to 2 FIB grids of the same thickness. Handy FIB grid holder on a standard 12.7 mm ( 1/2") pin stub. Can also be used to safely store FIB grids with thin sections attached. Overall Dimensions: 12.7 mm ( 1/2") x 7.8 mm (0.3 mm) H w/o pin. Pin: Standard 3.2 mm ( 1/8"). Material: Vacuum grade aluminum with brass screw. Includes Philips screwdriver #0.
Catalog No. 50-192-7127 Supplier Electron Microscopy Sciences Supplier No. 7595002
Holds up to 2 FIB grids of the same thickness. Handy FIB grid holder on a standard 12.7 mm ( 1/2") pin stub. Can also be used to safely store FIB grids with thin sections attached. Overall Dimensions: 12.7 mm ( 1/2") x 7.8 mm (0.3 mm) H w/o pin. Pin: Standard 3.2 mm ( 1/8"). Material: Vacuum grade aluminum with brass screw. Includes Philips screwdriver #0.