Learn More
Gilson Company Inc Small Fine Sieve Cleaning Brush
Catalog No. NC9034546
Encompass_Preferred
Small Fine Sieve Cleaning Brush with wood handle has soft, 100% China bristles in a round 0.75in (19mm) ferrule that is tapered for easy use with fine mesh sieves. This brush works well with 3in (76.2mm) or other small diameter sieves and Precision Electroformed sieves. The overall length is 5in (127mm).
Catalog No. NC9034546 Supplier Gilson Company Inc Supplier No. TSA168
Small Fine Sieve Cleaning Brush with wood handle has soft, 100% China bristles in a round 0.75in (19mm) ferrule that is tapered for easy use with fine mesh sieves. This brush works well with 3in (76.2mm) or other small diameter sieves and Precision Electroformed sieves. The overall length is 5in (127mm).