Learn More
Cole-Parmer Digi-Sense Small Fle x Thermistor, PVC Ins Sheath and Tip, Phono Plug, 13ft Lead
Catalog No. 501961496 Shop All Cole-Parmer Products
Encompass_Preferred
Note: Time constant is the time required to reach 63.2% of the new reading. To determine how long it will take to reach 99% of the new reading multiply the time constant by five. Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220F (104C); PVC handles are rated to 200F (93C); Stainless Steel handles are rated at 450F (232C); Mini Connectors are rated to 392F (200C).
Catalog No. 50-196-1496 Supplier Cole-Parmer Supplier No. 0849105
Note: Time constant is the time required to reach 63.2% of the new reading. To determine how long it will take to reach 99% of the new reading multiply the time constant by five. Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220F (104C); PVC handles are rated to 200F (93C); Stainless Steel handles are rated at 450F (232C); Mini Connectors are rated to 392F (200C).