Learn More
Saf T Pak Inc STP-309 - Replacement Outer Box and Insulated Chest only for STP 309SYS
SDP
Catalog No. NC9301949 Shop All Saf-T-Pak Inc. Products
Encompass_Preferred
STP-309 - Replacement Outer Box and Insulated Chest only for STP 309SYS, Diagnostic Shipper, Refrigerated; Cooler inside: 8 x 6 x 5in.; Outer box: 11 x 9 x 8in.; Lightweight packaging; For shipping diagnostic specimens require controlled tempature range, 8/Case
Catalog No. NC9301949 Supplier Saf T Pak Inc Supplier No. STP309
STP-309 - Replacement Outer Box and Insulated Chest only for STP 309SYS, Diagnostic Shipper, Refrigerated; Cooler inside: 8 x 6 x 5in.; Outer box: 11 x 9 x 8in.; Lightweight packaging; For shipping diagnostic specimens require controlled tempature range, 8/Case