Learn More
Saf T Pak Inc STP-309SYS Category B / Exempt Frozen Insulated Shipping System (UN 3373)
SDP
Catalog No. NC9018304 Shop All Saf-T-Pak Inc. Products
Encompass_Preferred
STP-309SYS Category B / Exempt Frozen Insulated Shipping System (UN 3373), Outer Box – OD 11.25 x 9.5 x 8.5in., pre-printed with Biological Substance, Category B and UN 3373 Markings, Insulated Chest – ID 7.75 x 5.75 x 5in., 8/Case
Catalog No. NC9018304 Supplier Saf T Pak Inc Supplier No. STP309SYS
STP-309SYS Category B / Exempt Frozen Insulated Shipping System (UN 3373), Outer Box – OD 11.25 x 9.5 x 8.5in., pre-printed with Biological Substance, Category B and UN 3373 Markings, Insulated Chest – ID 7.75 x 5.75 x 5in., 8/Case