Learn More
Promocell GmbH PromoCell Smooth Muscle Cell Basal Medium 2 (prf) 500 ml, phenol red-free
Catalog No. 50306209
Encompass
The PromoCell Smooth Muscle Cell Growth Medium 2 is a low-serum (5% V/V) medium developed to establish and maintain smooth muscle cell cultures from blood vessels and hollow organs.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, porcine, murine, dog, and rat smooth muscle cells.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.05 ml / ml || Epidermal Growth Factor (recombinant human) | 0.5 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 2 ng / ml || Insulin (recombinant human) | 5 μg / ml |
Catalog No. 50-306-209 Supplier Promocell GmbH Supplier No. C22267
The PromoCell Smooth Muscle Cell Growth Medium 2 is a low-serum (5% V/V) medium developed to establish and maintain smooth muscle cell cultures from blood vessels and hollow organs.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, porcine, murine, dog, and rat smooth muscle cells.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.05 ml / ml || Epidermal Growth Factor (recombinant human) | 0.5 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 2 ng / ml || Insulin (recombinant human) | 5 μg / ml |