Learn More
Promocell GmbH PromoCell Smooth Muscle Cell Growth Medium 2 500 ml, ready-to-use, low serum, with Supplement Mix C-39267 attached to the flask
Supplier:  Promocell GmbH C22062
Encompass_Preferred
The PromoCell Smooth Muscle Cell Growth Medium 2 is a low-serum (5% V/V) medium developed to establish and maintain smooth muscle cell cultures from blood vessels and hollow organs.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, porcine, murine, dog, and rat smooth muscle cells.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.05 ml / ml || Epidermal Growth Factor (recombinant human) | 0.5 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 2 ng / ml || Insulin (recombinant human) | 5 μg / ml |
Catalog No. 50-306-192