Learn More
Medchemexpress LLC HY-136848 5mg , SM1-71 CAS:2088179-99-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1368485MG
Encompass_Preferred
HY-136848 5mg SM1-71 CAS: 2088179-99-9 SM1-71 (compound 5) is a potent TAK1 inhibitor, with a Ki of 160 nM, it also can covalently inhibit MKNK2, MAP2K1/2/3/4/6/7, GAK, AAK1, BMP2K, MAP3K7, MAPKAPK5, GSK3A/B, MAPK1/3, SRC, YES1, FGFR1, ZAK (MLTK), MAP3K1, LIMK1 and RSK2. SM1-71 can inhibit proliferation of multiple cancer cell lines. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1411